Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCGGGGAGGCGGAGGACTCGGAG[C/G]GGGAAGAAGAGGAAGAGGAGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611205 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC88B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC88B - coiled-coil domain containing 88B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032251.5 | 168 | Missense Mutation | CGG,GGG | R,G 36 | NP_115627.6 | |
XM_006718519.3 | 168 | Missense Mutation | CGG,GGG | R,G 36 | XP_006718582.1 | |
XM_011544957.2 | 168 | Missense Mutation | CGG,GGG | R,G 36 | XP_011543259.1 | |
XM_011544959.2 | 168 | Missense Mutation | CGG,GGG | R,G 36 | XP_011543261.1 | |
XM_011544960.2 | 168 | Missense Mutation | CGG,GGG | R,G 36 | XP_011543262.1 | |
XM_011544962.2 | 168 | Intron | XP_011543264.1 | |||
XM_017017601.1 | 168 | Missense Mutation | CGG,GGG | R,G 36 | XP_016873090.1 |
MIR7155 - microRNA 7155 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |