Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACTTCTTCTCGCTTGTGCATGG[C/T]CTGACTCTGGTGGGCACGTCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609914 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AQP11 - aquaporin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173039.2 | 609 | Silent Mutation | GGC,GGT | G,G 90 | NP_766627.1 | |
XM_005273917.4 | 609 | Silent Mutation | GGC,GGT | G,G 90 | XP_005273974.1 |
LOC107987163 - uncharacterized LOC107987163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017018644.1 | 609 | Intron | XP_016874133.1 |