Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGTAGTGGCCCTTCCGCAGCAG[C/T]CGGTGTACACGGCCCACTGGGAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191350 MIM: 601772 MIM: 609806 | ||||||||||||||||||||
Literature Links: |
DPAGT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPAGT1 - dolichyl-phosphate N-acetylglucosaminephosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
H2AFX - H2A histone family member X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002105.2 | 172 | Silent Mutation | CGA,CGG | R,R 33 | NP_002096.1 |
HMBS - hydroxymethylbilane synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |