Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTTCTCCTCCAGCTCAGTCAGT[A/G]CCCAAGAGGTTTTCTGAAGCCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606096 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGALS12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGALS12 - galectin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142535.1 | Intron | NP_001136007.1 | ||||
NM_001142536.1 | Intron | NP_001136008.1 | ||||
NM_001142537.1 | Intron | NP_001136009.1 | ||||
NM_001142538.1 | Intron | NP_001136010.1 | ||||
NM_033101.3 | Intron | NP_149092.2 | ||||
XM_017018458.1 | Intron | XP_016873947.1 |