Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACACACCCCAAGGAGGCCAGCAC[A/G]GTGAGGTACTGGTTGATCTGCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 134690 MIM: 606866 MIM: 608046 | ||||||||||||||||||||
Literature Links: |
FAU PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAU - FAU, ubiquitin like and ribosomal protein S30 fusion | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6751 - microRNA 6751 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL49 - mitochondrial ribosomal protein L49 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYVN1 - synoviolin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032431.2 | 2520 | Silent Mutation | NP_115807.1 | |||
NM_172230.2 | 2520 | Silent Mutation | NP_757385.1 | |||
XM_011545303.2 | 2520 | Silent Mutation | XP_011543605.1 |