Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGATGGAGGCGCAACATGTTAC[A/G]CTGTGCTTGACAGAGGCAGTCACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603433 | ||||||||||||||||||||
Literature Links: |
ZNF143 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF143 - zinc finger protein 143 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282656.1 | 200 | Silent Mutation | ACA,ACG | T,T 27 | NP_001269585.1 | |
NM_001282657.1 | 200 | Silent Mutation | ACA,ACG | T,T 27 | NP_001269586.1 | |
NM_003442.5 | 200 | Silent Mutation | ACA,ACG | T,T 27 | NP_003433.3 | |
XM_005253121.1 | 200 | Silent Mutation | ACA,ACG | T,T 42 | XP_005253178.1 | |
XM_005253122.1 | 200 | Silent Mutation | ACA,ACG | T,T 42 | XP_005253179.1 | |
XM_005253127.1 | 200 | UTR 5 | XP_005253184.1 | |||
XM_011520349.2 | 200 | Silent Mutation | ACA,ACG | T,T 27 | XP_011518651.1 | |
XM_017018252.1 | 200 | Silent Mutation | ACA,ACG | T,T 42 | XP_016873741.1 | |
XM_017018253.1 | 200 | Silent Mutation | ACA,ACG | T,T 42 | XP_016873742.1 | |
XM_017018254.1 | 200 | Silent Mutation | ACA,ACG | T,T 27 | XP_016873743.1 | |
XM_017018255.1 | 200 | Silent Mutation | ACA,ACG | T,T 27 | XP_016873744.1 | |
XM_017018256.1 | 200 | Silent Mutation | ACA,ACG | T,T 27 | XP_016873745.1 |