Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGGCACTGTCAGAGGAGGAGC[A/G]GCTGAAGTTGGCTCAGCAGCAGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608234 MIM: 605591 | ||||||||||||||||||||
Literature Links: |
GAL3ST3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAL3ST3 - galactose-3-O-sulfotransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SF3B2 - splicing factor 3b subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006842.2 | 1050 | Missense Mutation | CAG,CGG | Q,R 146 | NP_006833.2 | |
XM_005273726.3 | 1050 | Missense Mutation | CAG,CGG | Q,R 146 | XP_005273783.1 | |
XM_011544740.2 | 1050 | Missense Mutation | CAG,CGG | Q,R 146 | XP_011543042.1 | |
XM_017017144.1 | 1050 | Missense Mutation | CAG,CGG | Q,R 146 | XP_016872633.1 |