Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTTTAAGAAACAAAGATAGTTT[G/T]CTGAACATTCTGTGTCCTGCCTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615936 | ||||||||||||||||||||
Literature Links: |
ARHGAP42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP42 - Rho GTPase activating protein 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152432.2 | 436 | Intron | NP_689645.2 | |||
XM_011542615.2 | 436 | UTR 3 | XP_011540917.1 | |||
XM_011542616.2 | 436 | UTR 3 | XP_011540918.1 | |||
XM_011542617.2 | 436 | UTR 3 | XP_011540919.1 | |||
XM_017017237.1 | 436 | UTR 3 | XP_016872726.1 | |||
XM_017017238.1 | 436 | UTR 3 | XP_016872727.1 |
TMEM133 - transmembrane protein 133 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032021.2 | 436 | Missense Mutation | TTG,TTT | L,F 69 | NP_114410.1 |