Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCGAACAGCAGCCCCGTGAGCA[C/T]ACTGAGGATCATTCTGGGGGTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611288 MIM: 612565 MIM: 611484 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNIH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNIH2 - cornichon family AMPA receptor auxiliary protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182553.2 | 813 | Intron | NP_872359.1 |
RAB1B - RAB1B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM151A - transmembrane protein 151A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YIF1A - Yip1 interacting factor homolog A, membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300861.1 | 813 | Missense Mutation | ATG,GTG | M,V 167 | NP_001287790.1 | |
NM_020470.2 | 813 | Missense Mutation | ATG,GTG | M,V 219 | NP_065203.2 | |
XM_005273720.3 | 813 | Missense Mutation | ATG,GTG | M,V 219 | XP_005273777.1 | |
XM_017017139.1 | 813 | Missense Mutation | ATG,GTG | M,V 231 | XP_016872628.1 | |
XM_017017140.1 | 813 | Missense Mutation | ATG,GTG | M,V 231 | XP_016872629.1 | |
XM_017017141.1 | 813 | Intron | XP_016872630.1 |