Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGCTGAAGTACTTTAGCCACAC[A/G]CGGAAGAACAGCCCACTACATTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM45B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM45B - transmembrane protein 45B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138788.3 | 329 | Silent Mutation | ACA,ACG | T,T 33 | NP_620143.1 | |
XM_005271400.4 | 329 | Silent Mutation | ACA,ACG | T,T 33 | XP_005271457.1 | |
XM_011542586.2 | 329 | Silent Mutation | ACA,ACG | T,T 33 | XP_011540888.1 | |
XM_011542588.2 | 329 | Silent Mutation | ACA,ACG | T,T 33 | XP_011540890.1 | |
XM_017017188.1 | 329 | Silent Mutation | ACA,ACG | T,T 33 | XP_016872677.1 | |
XM_017017189.1 | 329 | Silent Mutation | ACA,ACG | T,T 33 | XP_016872678.1 |