Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATGGGCCAGGACTGAGCTTCTC[A/G]ATGTCTGCATGAACGCCAAGCACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 136430 | ||||||||||||||||||||
Literature Links: |
FOLR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOLR1 - folate receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000802.3 | 148 | Missense Mutation | AAT,GAT | N,D 35 | NP_000793.1 | |
NM_016724.2 | 148 | Missense Mutation | AAT,GAT | N,D 35 | NP_057936.1 | |
NM_016725.2 | 148 | Missense Mutation | AAT,GAT | N,D 35 | NP_057937.1 | |
NM_016729.2 | 148 | Missense Mutation | AAT,GAT | N,D 35 | NP_057941.1 |