Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAATGTTTTTTATAGATCATCAC[C/T]GACCGACCTGGCTTTCATGATGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608422 MIM: 610079 MIM: 610614 | ||||||||||||||||||||
Literature Links: |
PANX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PANX3 - pannexin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIAE - sialic acid acetylesterase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBRG1 - transforming growth factor beta regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032811.2 | 644 | Silent Mutation | ACC,ACT | T,T 200 | NP_116200.2 | |
XM_005271709.3 | 644 | Silent Mutation | ACC,ACT | T,T 53 | XP_005271766.1 | |
XM_017018442.1 | 644 | Silent Mutation | ACC,ACT | T,T 49 | XP_016873931.1 |