Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGAAGACCTCCCAAAAATTGGTT[A/G]GTTAATTTGTAATGGTCCCAGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612880 | ||||||||||||||||||||
Literature Links: |
CCDC89 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC89 - coiled-coil domain containing 89 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYTL2 - synaptotagmin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162951.2 | 6602 | Silent Mutation | ACC,ACT | T,T 873 | NP_001156423.1 | |
NM_001162952.2 | 6602 | Silent Mutation | ACC,ACT | T,T 275 | NP_001156424.1 | |
NM_001162953.2 | 6602 | Silent Mutation | ACC,ACT | T,T 874 | NP_001156425.1 | |
NM_001289608.1 | 6602 | Silent Mutation | ACC,ACT | T,T 841 | NP_001276537.1 | |
NM_001289609.1 | 6602 | Silent Mutation | ACC,ACT | T,T 194 | NP_001276538.1 | |
NM_001289610.1 | 6602 | Silent Mutation | ACC,ACT | T,T 275 | NP_001276539.1 | |
NM_032943.4 | 6602 | Silent Mutation | ACC,ACT | T,T 849 | NP_116561.1 | |
NM_206929.3 | 6602 | Silent Mutation | ACC,ACT | T,T 315 | NP_996812.1 | |
NM_206930.3 | 6602 | Silent Mutation | ACC,ACT | T,T 304 | NP_996813.1 | |
XM_005274057.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2162 | XP_005274114.1 | |
XM_005274060.1 | 6602 | Silent Mutation | ACC,ACT | T,T 890 | XP_005274117.1 | |
XM_005274061.1 | 6602 | Silent Mutation | ACC,ACT | T,T 889 | XP_005274118.1 | |
XM_005274062.2 | 6602 | Silent Mutation | ACC,ACT | T,T 863 | XP_005274119.1 | |
XM_005274063.1 | 6602 | Silent Mutation | ACC,ACT | T,T 850 | XP_005274120.1 | |
XM_005274066.1 | 6602 | Silent Mutation | ACC,ACT | T,T 833 | XP_005274123.1 | |
XM_005274067.4 | 6602 | Silent Mutation | ACC,ACT | T,T 315 | XP_005274124.1 | |
XM_011545105.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2178 | XP_011543407.1 | |
XM_011545106.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2177 | XP_011543408.1 | |
XM_011545107.2 | 6602 | Silent Mutation | ACC,ACT | T,T 2151 | XP_011543409.1 | |
XM_011545108.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2138 | XP_011543410.1 | |
XM_011545109.2 | 6602 | Silent Mutation | ACC,ACT | T,T 2129 | XP_011543411.1 | |
XM_011545110.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2121 | XP_011543412.1 | |
XM_011545112.1 | 6602 | Intron | XP_011543414.1 | |||
XM_011545114.2 | 6602 | Silent Mutation | ACC,ACT | T,T 304 | XP_011543416.1 | |
XM_011545115.1 | 6602 | Silent Mutation | ACC,ACT | T,T 264 | XP_011543417.1 | |
XM_011545116.2 | 6602 | Silent Mutation | ACC,ACT | T,T 264 | XP_011543418.1 | |
XM_017017933.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2161 | XP_016873422.1 | |
XM_017017934.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2150 | XP_016873423.1 | |
XM_017017935.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2110 | XP_016873424.1 | |
XM_017017936.1 | 6602 | Silent Mutation | ACC,ACT | T,T 2035 | XP_016873425.1 | |
XM_017017937.1 | 6602 | Silent Mutation | ACC,ACT | T,T 815 | XP_016873426.1 | |
XM_017017938.1 | 6602 | Silent Mutation | ACC,ACT | T,T 801 | XP_016873427.1 | |
XM_017017939.1 | 6602 | Silent Mutation | ACC,ACT | T,T 384 | XP_016873428.1 | |
XM_017017940.1 | 6602 | Silent Mutation | ACC,ACT | T,T 368 | XP_016873429.1 |