Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTCCCAACCTCTTCTAACTCAT[C/T]GTCACAATGGGTTCCACAGATGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605569 | ||||||||||||||||||||
Literature Links: |
GPR83 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR83 - G protein-coupled receptor 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016540.3 | 1369 | Silent Mutation | ACA,ACG | T,T 421 | NP_057624.3 | |
XM_011542559.2 | 1369 | Silent Mutation | ACA,ACG | T,T 379 | XP_011540861.1 |