Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGCCCTCTCCCAGCACTGCTAC[A/G]CAGGTGAGTTCTGTGCAGGGAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605562 | ||||||||||||||||||||
Literature Links: |
SCGB2A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCGB2A2 - secretoglobin family 2A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002411.3 | 139 | Missense Mutation | ACA,GCA | T,A 18 | NP_002402.1 | |
XM_005274005.3 | 139 | Missense Mutation | ACA,GCA | T,A 18 | XP_005274062.3 |