Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGAATGGGCGGCCTCAACGTCCA[A/G]CTGGGCTCCTAGCACGTTCTCGTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602647 MIM: 603189 | ||||||||||||||||||||
Literature Links: |
NXF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NXF1 - nuclear RNA export factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX5 - syntaxin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244666.1 | 1094 | UTR 3 | NP_001231595.1 | |||
NM_003164.4 | 1094 | Silent Mutation | CTG,TTG | L,L 313 | NP_003155.2 | |
XM_011545223.2 | 1094 | Silent Mutation | CTG,TTG | L,L 259 | XP_011543525.1 | |
XM_011545224.2 | 1094 | Silent Mutation | CTG,TTG | L,L 217 | XP_011543526.1 |