Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGGACGAGGCCGCCTGGGAGGA[A/T]GAGGACGATGCAGATCTCCCCCACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605628 MIM: 603190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTATIP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTATIP2 - HIV-1 Tat interactive protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRMT3 - protein arginine methyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145166.1 | 322 | Intron | NP_001138638.1 | |||
NM_001145167.1 | 322 | Intron | NP_001138639.1 | |||
NM_005788.3 | 322 | Missense Mutation | GAA,GAT | E,D 35 | NP_005779.1 | |
XM_011519836.2 | 322 | Intron | XP_011518138.1 | |||
XM_017017081.1 | 322 | Missense Mutation | AAG,ATG | K,M 19 | XP_016872570.1 |