Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTTAAAATATGTGGGGACCCTC[C/T]CTCCATGACTGGGTTCCCCTGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC81 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC81 - coiled-coil domain containing 81 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001156474.1 | Intron | NP_001149946.1 | ||||
NM_021827.4 | Intron | NP_068599.3 | ||||
XM_005274161.1 | Intron | XP_005274218.1 | ||||
XM_005274162.3 | Intron | XP_005274219.1 | ||||
XM_017018105.1 | Intron | XP_016873594.1 |