Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTAATTGATTAACTTCTCCATA[A/G]TCATAGTCATCATACCTACGAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607768 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SESN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SESN3 - sestrin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271594.1 | 1589 | Silent Mutation | GAC,GAT | D,D 282 | NP_001258523.1 | |
NM_144665.3 | 1589 | Silent Mutation | GAC,GAT | D,D 421 | NP_653266.2 | |
XM_011542614.1 | 1589 | Silent Mutation | GAC,GAT | D,D 398 | XP_011540916.1 |