Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCGGCCAGGCGGGCCTGCAGCTC[A/G]CGGCTCAGCTCCTCGTGGTCCACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600856 MIM: 603240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKN1C - cyclin dependent kinase inhibitor 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000076.2 | 270 | Silent Mutation | CGC,CGT | R,R 44 | NP_000067.1 | |
NM_001122630.1 | 270 | Silent Mutation | CGC,CGT | R,R 33 | NP_001116102.1 | |
NM_001122631.1 | 270 | Silent Mutation | CGC,CGT | R,R 33 | NP_001116103.1 | |
XM_005252732.4 | 270 | Silent Mutation | CGC,CGT | R,R 33 | XP_005252789.1 | |
XM_017017088.1 | 270 | Silent Mutation | CGC,CGT | R,R 44 | XP_016872577.1 |
SLC22A18AS - solute carrier family 22 member 18 antisense | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |