Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAGCCTCCAGGGGCAGGACATG[A/G]AACAGGACGCGGCAGCCGGGCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603166 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP4K2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP4K2 - mitogen-activated protein kinase kinase kinase kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307990.1 | 1983 | Silent Mutation | TTC,TTT | F,F 672 | NP_001294919.1 | |
NM_004579.4 | 1983 | Silent Mutation | TTC,TTT | F,F 680 | NP_004570.2 | |
XM_011545203.2 | 1983 | Silent Mutation | TTC,TTT | F,F 680 | XP_011543505.1 | |
XM_011545204.2 | 1983 | Silent Mutation | TTC,TTT | F,F 636 | XP_011543506.1 | |
XM_017018093.1 | 1983 | Silent Mutation | TTC,TTT | F,F 628 | XP_016873582.1 | |
XM_017018094.1 | 1983 | Silent Mutation | TTC,TTT | F,F 503 | XP_016873583.1 | |
XM_017018095.1 | 1983 | Silent Mutation | TTC,TTT | F,F 503 | XP_016873584.1 | |
XM_017018096.1 | 1983 | Intron | XP_016873585.1 |