Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAATGATGGAGATCAGGGAACA[A/G]GCCTTGGGGATCCTGGAGCGGGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130593 MIM: 610641 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EEF1G - eukaryotic translation elongation factor 1 gamma | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6747 - microRNA 6747 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUT1 - terminal uridylyl transferase 1, U6 snRNA-specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022830.2 | 2712 | Silent Mutation | CTG,TTG | L,L 892 | NP_073741.2 |