Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCTCGAAGCACAAGGTGTGC[A/G]TGGACCTGAGGGCCAACCTGAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607719 MIM: 191043 | ||||||||||||||||||||
Literature Links: |
SYT8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYT8 - synaptotagmin 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNNI2 - troponin I2, fast skeletal type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145829.1 | 425 | Missense Mutation | ATG,GTG | M,V 135 | NP_001139301.1 | |
NM_001145841.1 | 425 | Missense Mutation | ATG,GTG | M,V 135 | NP_001139313.1 | |
NM_003282.3 | 425 | Missense Mutation | ATG,GTG | M,V 135 | NP_003273.1 |