Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTCCGCGCCGCGAGGAGGAGGCC[C/G]TGCTGGTTTCTGTGCGGGTGAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612409 MIM: 602571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBM14 - RNA binding motif protein 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM14-RBM4 - RBM14-RBM4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198845.1 | 119 | Intron | NP_001185774.1 | |||
NM_001198846.1 | 119 | Intron | NP_001185775.1 |
RBM4 - RNA binding motif protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198843.1 | 119 | UTR 5 | NP_001185772.1 | |||
NM_001198844.1 | 119 | UTR 5 | NP_001185773.1 | |||
NM_002896.3 | 119 | UTR 5 | NP_002887.2 |