Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCGCGGCAGCTCCATGGAAATCA[A/G]TCTGGGGGAGAAGGACACTGCATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601998 MIM: 605720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRRA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESRRA - estrogen related receptor alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNK4 - potassium two pore domain channel subfamily K member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317090.1 | 334 | Intron | NP_001304019.1 | |||
NM_033310.2 | 334 | Intron | NP_201567.1 |
KCNK4-TEX40 - KCNK4-TEX40 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX40 - testis expressed 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039496.1 | 334 | Missense Mutation | AAT,AGT | N,S 105 | NP_001034585.1 |