Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGTACCAGCCATCCAGACAGT[A/G]CTTGAGCTGCAGAAACTGAGACCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606158 MIM: 608941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BSCL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BSCL2 - BSCL2, seipin lipid droplet biogenesis associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122955.3 | 114 | Intron | NP_001116427.1 | |||
NM_001130702.2 | 114 | Intron | NP_001124174.2 | |||
NM_032667.6 | 114 | Intron | NP_116056.3 |
GNG3 - G protein subunit gamma 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012202.4 | 114 | UTR 5 | NP_036334.1 | |||
XM_006718500.2 | 114 | UTR 5 | XP_006718563.1 |
HNRNPUL2 - heterogeneous nuclear ribonucleoprotein U like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPUL2-BSCL2 - HNRNPUL2-BSCL2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |