Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCGTGCAGGTCTTCTCCAGAACA[A/T]CTTGGAAGAAGGTAATGACCTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603211 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC82 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC82 - coiled-coil domain containing 82 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
JRKL - JRK-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261833.1 | 1693 | Missense Mutation | AAC,ATC | N,I 346 | NP_001248762.1 | |
NM_003772.3 | 1693 | Missense Mutation | AAC,ATC | N,I 346 | NP_003763.2 |