Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCATTATTGTCATTTGTTACAG[G/T]AAAGTATTTTAACAGAGTAGCGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615251 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBED5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBED5 - zinc finger BED-type containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143667.1 | 2192 | Missense Mutation | ACT,CCT | T,P 565 | NP_001137139.1 | |
NM_021211.3 | 2192 | Missense Mutation | ACT,CCT | T,P 565 | NP_067034.2 |
ZBED5-AS1 - ZBED5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |