Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTATGTGTATATGTGCAAGGATGG[G/T]TATGCCGCCATGGCAGCACAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGBL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGBL2 - ATP/GTP binding protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024783.3 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 893 | NP_079059.2 | |
XM_005253138.4 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 855 | XP_005253195.1 | |
XM_005253139.4 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 819 | XP_005253196.1 | |
XM_005253140.4 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 777 | XP_005253197.1 | |
XM_005253141.4 | 2481 | Intron | XP_005253198.1 | |||
XM_006718327.3 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 837 | XP_006718390.1 | |
XM_006718328.3 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 801 | XP_006718391.1 | |
XM_011520372.2 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 855 | XP_011518674.1 | |
XM_011520373.2 | 2481 | Intron | XP_011518675.1 | |||
XM_011520374.2 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 819 | XP_011518676.1 | |
XM_011520375.2 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 810 | XP_011518677.1 | |
XM_011520376.2 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 757 | XP_011518678.1 | |
XM_011520377.2 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 757 | XP_011518679.1 | |
XM_011520378.2 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 757 | XP_011518680.1 | |
XM_011520379.2 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 757 | XP_011518681.1 | |
XM_011520381.2 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 748 | XP_011518683.1 | |
XM_017018331.1 | 2481 | Intron | XP_016873820.1 | |||
XM_017018332.1 | 2481 | Nonsense Mutation | TAA,TAC | *,Y 817 | XP_016873821.1 | |
XM_017018333.1 | 2481 | Intron | XP_016873822.1 |