Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGATGGTCTCCATGCTAGAAGAA[A/C]GGGACCGCCTTCTTGATACACTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611054 | ||||||||||||||||||||
Literature Links: |
PPFIA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPFIA1 - PTPRF interacting protein alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003626.3 | 367 | Silent Mutation | AGG,CGG | R,R 47 | NP_003617.1 | |
NM_177423.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | NP_803172.1 | |
XM_006718716.3 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_006718779.1 | |
XM_011545306.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543608.1 | |
XM_011545307.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543609.1 | |
XM_011545308.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543610.1 | |
XM_011545309.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543611.1 | |
XM_011545310.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543612.1 | |
XM_011545311.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543613.1 | |
XM_011545312.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543614.1 | |
XM_011545313.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543615.1 | |
XM_011545314.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543616.1 | |
XM_011545315.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543617.1 | |
XM_011545316.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543618.1 | |
XM_011545317.2 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543619.1 | |
XM_011545318.2 | 367 | Intron | XP_011543620.1 | |||
XM_011545319.1 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_011543621.1 | |
XM_017018448.1 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_016873937.1 | |
XM_017018449.1 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_016873938.1 | |
XM_017018450.1 | 367 | Silent Mutation | AGG,CGG | R,R 47 | XP_016873939.1 |