Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTGCTTTTTTCCTCTGATCCTC[A/C]AAGCTTTCTCGGATGCCTTTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CWF19L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CWF19L2 - CWF19-like 2, cell cycle control (S. pombe) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152434.2 | 2637 | Missense Mutation | TTG,TTT | L,F 869 | NP_689647.2 | |
XM_011542620.2 | 2637 | Intron | XP_011540922.1 | |||
XM_017017239.1 | 2637 | Missense Mutation | TTG,TTT | L,F 393 | XP_016872728.1 |
LOC105369477 - uncharacterized LOC105369477 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |