Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCTGACGTGCTGGTATACCTAG[C/T]GGATGACACGGTGGTGCCCCTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610825 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRMD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRMD8 - FERM domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300832.1 | Intron | NP_001287761.1 | ||||
NM_001300833.1 | Intron | NP_001287762.1 | ||||
NM_031904.3 | Intron | NP_114110.1 |
SLC25A45 - solute carrier family 25 member 45 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |