Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCATGATGCTGACGTCCCGGCT[A/G]GTCCGGCGCACCGCAGCACTCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609632 | ||||||||||||||||||||
Literature Links: |
GDPD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDPD5 - glycerophosphodiester phosphodiesterase domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030792.6 | 2895 | Silent Mutation | ACC,ACT | T,T 542 | NP_110419.5 | |
XM_006718697.3 | 2895 | Silent Mutation | ACC,ACT | T,T 542 | XP_006718760.1 | |
XM_011545276.2 | 2895 | Silent Mutation | ACC,ACT | T,T 542 | XP_011543578.1 | |
XM_011545277.2 | 2895 | Silent Mutation | ACC,ACT | T,T 542 | XP_011543579.1 | |
XM_011545278.2 | 2895 | Silent Mutation | ACC,ACT | T,T 542 | XP_011543580.1 | |
XM_011545279.2 | 2895 | Silent Mutation | ACC,ACT | T,T 542 | XP_011543581.1 | |
XM_011545280.1 | 2895 | Silent Mutation | ACC,ACT | T,T 485 | XP_011543582.1 | |
XM_011545281.2 | 2895 | Silent Mutation | ACC,ACT | T,T 404 | XP_011543583.1 | |
XM_011545285.2 | 2895 | Silent Mutation | ACC,ACT | T,T 404 | XP_011543587.1 | |
XM_011545286.1 | 2895 | Silent Mutation | ACC,ACT | T,T 297 | XP_011543588.1 | |
XM_011545287.1 | 2895 | Silent Mutation | ACC,ACT | T,T 297 | XP_011543589.1 |
KLHL35 - kelch like family member 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |