Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGCCATATCCACAGGGAAGGG[C/T]CTGCTGGGCTCCCATGCTGTGTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606374 MIM: 104160 MIM: 180721 | ||||||||||||||||||||
Literature Links: |
B3GAT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B3GAT3 - beta-1,3-glucuronyltransferase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288721.1 | 969 | Silent Mutation | AGA,AGG | R,R 240 | NP_001275650.1 | |
NM_001288722.1 | 969 | Silent Mutation | AGA,AGG | R,R 247 | NP_001275651.1 | |
NM_001288723.1 | 969 | Silent Mutation | AGA,AGG | R,R 247 | NP_001275652.1 | |
NM_012200.3 | 969 | Silent Mutation | AGA,AGG | R,R 247 | NP_036332.2 |
EML3 - echinoderm microtubule associated protein like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GANAB - glucosidase II alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ROM1 - retinal outer segment membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |