Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAGCTTGGCAGAGCGCTGGCGC[C/T]GTCGGGCCCGGCACTCCACGTGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607233 | ||||||||||||||||||||
Literature Links: |
MRGPRF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRGPRF - MAS related GPR family member F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098515.1 | 1554 | Missense Mutation | CAG,CGG | Q,R 230 | NP_001091985.1 | |
NM_145015.4 | 1554 | Missense Mutation | CAG,CGG | Q,R 230 | NP_659452.3 | |
XM_017017170.1 | 1554 | Missense Mutation | CAG,CGG | Q,R 230 | XP_016872659.1 |
MRGPRF-AS1 - MRGPRF antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |