Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGAGAGCTCTGTGAAATTCTCA[G/T]GTAGCTGAAACTTTTTCTCATCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609962 | ||||||||||||||||||||
Literature Links: |
CLEC4E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC4E - C-type lectin domain family 4 member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014358.3 | 344 | Missense Mutation | CAT,CCT | H,P 60 | NP_055173.1 | |
XM_011520614.2 | 344 | Missense Mutation | CAT,CCT | H,P 29 | XP_011518916.1 | |
XM_011520615.1 | 344 | Missense Mutation | CAT,CCT | H,P 60 | XP_011518917.1 |