Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGGGACGGGTCTACGGGGACG[C/G]GCTGCGGGACCTGTCCGGCTTGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601621 | ||||||||||||||||||||
Literature Links: |
TBX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBX3 - T-box 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005996.3 | 3125 | Missense Mutation | CCG,GCG | P,A 721 | NP_005987.3 | |
NM_016569.3 | 3125 | Missense Mutation | CCG,GCG | P,A 741 | NP_057653.3 |