Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTCCCATCTTCCTCTGACCCA[C/G]AGTCCAAGTCTGGGGAAGAATGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608870 | ||||||||||||||||||||
Literature Links: |
LRIG3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRIG3 - leucine rich repeats and immunoglobulin like domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136051.2 | 3539 | Missense Mutation | TCT,TGT | S,C 1019 | NP_001129523.1 | |
NM_153377.4 | 3539 | Missense Mutation | TCT,TGT | S,C 1079 | NP_700356.2 | |
XM_011537893.2 | 3539 | Missense Mutation | TCT,TGT | S,C 1001 | XP_011536195.1 | |
XM_017018790.1 | 3539 | Intron | XP_016874279.1 |