Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTGGGCTGCTGGTGATCACGGAG[A/G]GCTCCGGGCTCTGTATGGGCAGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609843 | ||||||||||||||||||||
Literature Links: |
DCP1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCP1B - decapping mRNA 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319292.1 | 1931 | Intron | NP_001306221.1 | |||
NM_152640.4 | 1931 | Missense Mutation | CCC,CTC | P,L 569 | NP_689853.3 | |
XM_011520927.2 | 1931 | Missense Mutation | CCC,CTC | P,L 467 | XP_011519229.1 |