Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTACAGTTTTGGTTAGCTGCAC[A/G]GCATTATCCTGTCTTAGAGATTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613481 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC62 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC62 - coiled-coil domain containing 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_201435.4 | Intron | NP_958843.2 | ||||
XM_006719643.1 | Intron | XP_006719706.1 | ||||
XM_006719644.1 | Intron | XP_006719707.1 | ||||
XM_011538856.1 | Intron | XP_011537158.1 | ||||
XM_011538857.1 | Intron | XP_011537159.1 | ||||
XM_011538859.1 | Intron | XP_011537161.1 | ||||
XM_017020033.1 | Intron | XP_016875522.1 |