Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAATGCTGGCCGCCAACCCAGG[A/C]AAGACCCCGATCAGCCTTCTGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606371 MIM: 600447 MIM: 604643 MIM: 605053 | ||||||||||||||||||||
Literature Links: |
ATF7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATF7 - activating transcription factor 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP3K12 - mitogen-activated protein kinase kinase kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193511.1 | 191 | Intron | NP_001180440.1 | |||
NM_006301.3 | 191 | Intron | NP_006292.3 | |||
XM_005269138.3 | 191 | Intron | XP_005269195.1 | |||
XM_005269140.2 | 191 | Intron | XP_005269197.1 | |||
XM_006719588.3 | 191 | Intron | XP_006719651.1 | |||
XM_011538725.2 | 191 | Intron | XP_011537027.1 | |||
XM_017019956.1 | 191 | Intron | XP_016875445.1 |
NPFF - neuropeptide FF-amide peptide precursor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARBP2 - TARBP2, RISC loading complex RNA binding subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004178.4 | 191 | Silent Mutation | GGA,GGC | G,G 7 | NP_004169.3 | |
NM_134323.1 | 191 | Silent Mutation | GGA,GGC | G,G 28 | NP_599150.1 | |
NM_134324.2 | 191 | Silent Mutation | GGA,GGC | G,G 7 | NP_599151.2 | |
XM_005269114.1 | 191 | Silent Mutation | GGA,GGC | G,G 7 | XP_005269171.1 | |
XM_005269115.2 | 191 | Silent Mutation | GGA,GGC | G,G 7 | XP_005269172.1 | |
XM_005269117.1 | 191 | Silent Mutation | GGA,GGC | G,G 28 | XP_005269174.1 | |
XM_005269120.4 | 191 | UTR 5 | XP_005269177.1 | |||
XM_005269122.3 | 191 | UTR 5 | XP_005269179.1 | |||
XM_006719581.1 | 191 | Intron | XP_006719644.1 | |||
XM_011538712.2 | 191 | UTR 5 | XP_011537014.1 | |||
XM_017019910.1 | 191 | Silent Mutation | GGA,GGC | G,G 7 | XP_016875399.1 | |
XM_017019911.1 | 191 | UTR 5 | XP_016875400.1 |