Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGACGGAGGCCCTGCCCTCAGA[C/G]ACGCGCCCCCTAGTGGAAGCAGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC59 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC59 - coiled-coil domain containing 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014167.4 | 294 | Intron | NP_054886.2 |
METTL25 - methyltransferase like 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319675.1 | 294 | UTR 5 | NP_001306604.1 | |||
NM_032230.2 | 294 | Missense Mutation | GAC,GAG | D,E 75 | NP_115606.2 | |
XM_005269186.4 | 294 | Missense Mutation | GAC,GAG | D,E 75 | XP_005269243.2 | |
XM_011538827.2 | 294 | Missense Mutation | GAC,GAG | D,E 75 | XP_011537129.1 | |
XM_011538828.2 | 294 | Missense Mutation | GAC,GAG | D,E 75 | XP_011537130.1 | |
XM_011538829.2 | 294 | Missense Mutation | GAC,GAG | D,E 75 | XP_011537131.1 | |
XM_017020020.1 | 294 | UTR 5 | XP_016875509.1 | |||
XM_017020021.1 | 294 | UTR 5 | XP_016875510.1 |