Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTGGGAAATTCGCATTTATTC[A/G]TGAATCGTTCCAACAAACTTGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601750 | ||||||||||||||||||||
Literature Links: |
GTF2H3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTF2H3 - general transcription factor IIH subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271866.1 | 272 | Missense Mutation | ATG,GTG | M,V 51 | NP_001258795.1 | |
NM_001271867.1 | 272 | Missense Mutation | ATG,GTG | M,V 10 | NP_001258796.1 | |
NM_001271868.1 | 272 | UTR 5 | NP_001258797.1 | |||
NM_001516.4 | 272 | Missense Mutation | ATG,GTG | M,V 51 | NP_001507.2 | |
XM_017019228.1 | 272 | UTR 5 | XP_016874717.1 |