Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGGGGTGGCGTGGTGGATCCGA[C/T]GTGCCACCCCGACGTGGGCCTTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605884 | ||||||||||||||||||||
Literature Links: |
CCDC92 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC92 - coiled-coil domain containing 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304957.1 | 1302 | Missense Mutation | CAT,CGT | H,R 293 | NP_001291886.1 | |
NM_001304958.1 | 1302 | Missense Mutation | CAT,CGT | H,R 293 | NP_001291887.1 | |
NM_001304959.1 | 1302 | Missense Mutation | CAT,CGT | H,R 293 | NP_001291888.1 | |
NM_001304960.1 | 1302 | Missense Mutation | CAT,CGT | H,R 293 | NP_001291889.1 | |
NM_001304961.1 | 1302 | Missense Mutation | CAT,CGT | H,R 276 | NP_001291890.1 | |
NM_025140.2 | 1302 | Missense Mutation | CAT,CGT | H,R 293 | NP_079416.1 | |
XM_005253624.2 | 1302 | Missense Mutation | CAT,CGT | H,R 293 | XP_005253681.1 | |
XM_017019984.1 | 1302 | Missense Mutation | CAT,CGT | H,R 293 | XP_016875473.1 |
DNAH10 - dynein axonemal heavy chain 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |