Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTTATGGTGGCGCGGCGGACTC[C/G]GTGCGCTCCTCCAACTTGTCTTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
BTBD11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTBD11 - BTB domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017523.1 | 1857 | Intron | NP_001017523.1 | |||
NM_001018072.1 | 1857 | Silent Mutation | TCC,TCG | S,S 28 | NP_001018082.1 | |
XM_005268645.3 | 1857 | Intron | XP_005268702.1 | |||
XM_011537909.2 | 1857 | Intron | XP_011536211.1 | |||
XM_011537910.2 | 1857 | Intron | XP_011536212.1 | |||
XM_017018807.1 | 1857 | Silent Mutation | TCC,TCG | S,S 28 | XP_016874296.1 |