Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATACAGAAAAAGTATTAACAAAC[A/G]GAGAAGCCCCAGATACACGTACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615806 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC15A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC15A4 - solute carrier family 15 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145648.3 | 2828 | UTR 3 | NP_663623.1 | |||
XM_011537895.1 | 2828 | UTR 3 | XP_011536197.1 | |||
XM_017018791.1 | 2828 | Intron | XP_016874280.1 | |||
XM_017018792.1 | 2828 | Intron | XP_016874281.1 | |||
XM_017018793.1 | 2828 | Intron | XP_016874282.1 |