Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTGTTCTTCATTGTCTTTACTT[A/G]TAAGGTGTTGAAGGTGGGATCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614530 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFA12 - NADH:ubiquinone oxidoreductase subunit A12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258338.1 | 377 | UTR 3 | NP_001245267.1 | |||
NM_018838.4 | 377 | Silent Mutation | TAC,TAT | Y,Y 144 | NP_061326.1 |