Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAGAGCTGGCGTCCAAGCTGGA[C/T]CACTACAAAGCCAAGGCCACGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614939 | ||||||||||||||||||||
Literature Links: |
PGAM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGAM5 - PGAM family member 5, mitochondrial serine/threonine protein phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170543.1 | 340 | Silent Mutation | GAC,GAT | D,D 90 | NP_001164014.1 | |
NM_001170544.1 | 340 | Silent Mutation | GAC,GAT | D,D 90 | NP_001164015.1 | |
NM_138575.3 | 340 | Silent Mutation | GAC,GAT | D,D 90 | NP_612642.2 |
PXMP2 - peroxisomal membrane protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |