Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATGATTGTCCCATACCACATAGA[C/T]GCTGCCACTGTCCCCCTGGCAGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613785 MIM: 608974 | ||||||||||||||||||||
Literature Links: |
C1R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1R - complement C1r subcomponent | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1RL - complement C1r subcomponent like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297640.1 | 1460 | Missense Mutation | NP_001284569.1 | |||
NM_001297642.1 | 1460 | UTR 3 | NP_001284571.1 | |||
NM_001297643.1 | 1460 | Intron | NP_001284572.1 | |||
NM_016546.3 | 1460 | Missense Mutation | NP_057630.2 | |||
XM_017019397.1 | 1460 | Missense Mutation | XP_016874886.1 | |||
XM_017019398.1 | 1460 | Missense Mutation | XP_016874887.1 | |||
XM_017019399.1 | 1460 | Intron | XP_016874888.1 | |||
XM_017019400.1 | 1460 | Intron | XP_016874889.1 | |||
XM_017019401.1 | 1460 | Intron | XP_016874890.1 | |||
XM_017019402.1 | 1460 | Intron | XP_016874891.1 | |||
XM_017019403.1 | 1460 | Intron | XP_016874892.1 | |||
XM_017019404.1 | 1460 | Intron | XP_016874893.1 |