Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCCCCGCCCCTTGACTGTTCTC[G/T]CGTTCGCGGACGGCTGTGGTGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609695 MIM: 603146 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPD - 4-hydroxyphenylpyruvate dioxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171993.1 | 11 | Intron | NP_001165464.1 | |||
NM_002150.2 | 11 | Intron | NP_002141.1 |
PSMD9 - proteasome 26S subunit, non-ATPase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261400.2 | 11 | UTR 5 | NP_001248329.1 | |||
NM_002813.6 | 11 | UTR 5 | NP_002804.2 |